Learn More
Electron Microscopy Sciences CryoFlex Tubing (1.4 x 50 cm) 15/pk
SDP
Catalog No. 501899010
Encompass_Preferred
CryoFlex is a hollow tube from polyethylene which is useful for encasing and sealing cryo vials, when storing samples immersed in liquid nitrogen. The tubing measures 9/16" diameter x 1911/16" long (1.4 x 50cm)
Catalog No. 50-189-9010 Supplier Electron Microscopy Sciences Supplier No. 6108310
CryoFlex is a hollow tube from polyethylene which is useful for encasing and sealing cryo vials, when storing samples immersed in liquid nitrogen. The tubing measures 9/16" diameter x 1911/16" long (1.4 x 50cm)