Learn More
Research Products International Corp Cryo-Freeze Storage Boxes, 25 Tube Capacity, Assorted Colors, 5 per Package, 3 x 3 x 2 1/8
SDP
Catalog No. 50476467
Encompass
Cryo-Freeze boxes are designed for storage at ultra-low cryogenic temperatures. Manufactured from high strength polycarbonate for maximum durability and performance at sub-zero temperature conditions. Each box features a permanent numeric grid index, vented lid and base allows liquid drainage and eliminates any pressure differential. Accommodate all standard micro-tubes and cryogenic vials.
Catalog No. 50-476-467 Supplier Research Products International Corp Supplier No. 145925
Cryo-Freeze boxes are designed for storage at ultra-low cryogenic temperatures. Manufactured from high strength polycarbonate for maximum durability and performance at sub-zero temperature conditions. Each box features a permanent numeric grid index, vented lid and base allows liquid drainage and eliminates any pressure differential. Accommodate all standard micro-tubes and cryogenic vials.