Learn More
Research Products International Corp Cryo-Freeze Storage Boxes, 81 Tube Capacity, Assorted Colors, 5 per Package
SDP
Catalog No. 50476468
Encompass
Cryo-Freeze boxes are designed for storage at ultra-low cryogenic temperatures. Manufactured from high strength polycarbonate for maximum durability and performance at sub-zero temperature conditions. Each box features a permanent numeric grid index, vented lid and base allows liquid drainage and eliminates any pressure differential. Accommodate all standard micro-tubes and cryogenic vials.
Catalog No. 50-476-468 Supplier Research Products International Corp Supplier No. 145981
Cryo-Freeze boxes are designed for storage at ultra-low cryogenic temperatures. Manufactured from high strength polycarbonate for maximum durability and performance at sub-zero temperature conditions. Each box features a permanent numeric grid index, vented lid and base allows liquid drainage and eliminates any pressure differential. Accommodate all standard micro-tubes and cryogenic vials.