Learn More
Electron Microscopy Sciences CryoELITE Cryogenic Vials, Blue, 2 mL Volume
SDP
Catalog No. 501929353
Encompass_Preferred
CryoELITE cryogenic vials feature unrivaled cap seal that exceeds DOT and IATA regulations ensuring ultimate protection of samples during transportation and demanding freeze-thaw handling. Made from low binding, cryogenic grade virgin polypropylene.
Lot certified Rnase/Dnase and endotoxin Free providing assurance of product integrity.
Conical well design allows for precise sample retrieval from both automated and manual systems.
Loctagon Vial Skirt provides stability in freestanding position and locks into CryoELITE Benchmate Vial Rack in order to provide easy open and close with one hand.
Colored cap allows for color coding projects along with WHEATON colored freezer and storage boxes. Features writing patch. Sterile.
Color: Blue
Volume: 2 mL
Catalog No. 50-192-9353 Supplier Electron Microscopy Sciences Supplier No. 61807B
CryoELITE cryogenic vials feature unrivaled cap seal that exceeds DOT and IATA regulations ensuring ultimate protection of samples during transportation and demanding freeze-thaw handling. Made from low binding, cryogenic grade virgin polypropylene.
Lot certified Rnase/Dnase and endotoxin Free providing assurance of product integrity.
Conical well design allows for precise sample retrieval from both automated and manual systems.
Loctagon Vial Skirt provides stability in freestanding position and locks into CryoELITE Benchmate Vial Rack in order to provide easy open and close with one hand.
Colored cap allows for color coding projects along with WHEATON colored freezer and storage boxes. Features writing patch. Sterile.
Color: Blue
Volume: 2 mL