Learn More
CELLTREAT Scientific Products 40 Place Easy-Grip Workstation Rack for CF Cryogenic Vial, Non-sterile
SDP
Catalog No. 501149099
Encompass_Preferred
CELLTREAT Scientific Products Easy-Grip Workstation Rack. Easy-Grip Workstation Rack has individual positions which feature ribs to interlock with self-standing vials for easy one hand opening, features four gripping feet for increased stability and reduced slipping, and has low-profile design for maximum storage efficiency. Holds 40 Freeze Cryogenic Vials. Labeled with alpha-numeric markings for easy identification of individual positions. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 2/re-sealable bag, 2/case.
Catalog No. 50-114-9099 Supplier CELLTREAT Scientific Products Supplier No. 229941
CELLTREAT Scientific Products Easy-Grip Workstation Rack. Easy-Grip Workstation Rack has individual positions which feature ribs to interlock with self-standing vials for easy one hand opening, features four gripping feet for increased stability and reduced slipping, and has low-profile design for maximum storage efficiency. Holds 40 Freeze Cryogenic Vials. Labeled with alpha-numeric markings for easy identification of individual positions. RNase/DNase free and non-pyrogenic. Every case is printed with the lot number for quality traceability. Packaged non-sterile, 2/re-sealable bag, 2/case.