Learn More
Electron Microscopy Sciences Cryogenic Cardboard Box and Lid for 15 and 50 ml tubes, 145 x 145 x 122 mm
SDP
Catalog No. NC1164602
Encompass_Preferred
Low-cost choice with excellent low-temperature storage properties. One-size box safely stores 15 and 50 ml tubes in LN2 or mechanical freezers.
Catalog No. NC1164602 Supplier Electron Microscopy Sciences Supplier No. 41005
Low-cost choice with excellent low-temperature storage properties. One-size box safely stores 15 and 50 ml tubes in LN2 or mechanical freezers.