Learn More
GA International Cryogenic Barcode Labels for Liquid Nitrogen Storage 1" x 0.5" / 25.4mm x 12.7mm Rectangle Blue From -196°C to +110°C (-321°F to +230°F)
Catalog No. 5000571215
Encompass_Preferred
NitroTAG labels are available in a range of sizes and configurations, perfect for identifying small-diameter cryo vials, microtubes and tube tops, cardboard or polypropylene cryo/freezer boxes, cell culture plates, and plastic bags.These thermal-transfer cryogenic labels have a strong permanent adhesive that will remain firmly and permanently attached in ultra-low temperatures. They are designed for long-term cryo-preservation under cryogenic laboratory conditions, including in liquid and vapor phase liquid nitrogen (-196°C/-321°F), freezers (-80°C/-112°F), and dry ice transportation.NitroTAG labels require zero cure time and can be immersed in liquid nitrogen immediately after application without the need for additional tape or lamination. Additionally, these labels are waterproof, weatherproof, and durable. NitroTAG labels will adhere to wet surfaces, resist multiple freeze-thaw cycles, and can withstand elevated temperatures up to +110°C.They are also resistant to spraying wi
Catalog No. 50-005-71215 Supplier GA International Supplier No. JTTA7C35BL
NitroTAG labels are available in a range of sizes and configurations, perfect for identifying small-diameter cryo vials, microtubes and tube tops, cardboard or polypropylene cryo/freezer boxes, cell culture plates, and plastic bags.These thermal-transfer cryogenic labels have a strong permanent adhesive that will remain firmly and permanently attached in ultra-low temperatures. They are designed for long-term cryo-preservation under cryogenic laboratory conditions, including in liquid and vapor phase liquid nitrogen (-196°C/-321°F), freezers (-80°C/-112°F), and dry ice transportation.NitroTAG labels require zero cure time and can be immersed in liquid nitrogen immediately after application without the need for additional tape or lamination. Additionally, these labels are waterproof, weatherproof, and durable. NitroTAG labels will adhere to wet surfaces, resist multiple freeze-thaw cycles, and can withstand elevated temperatures up to +110°C.They are also resistant to spraying wi