Learn More
Alkali Scientific ASI™ Cryogenic Boxes with Hinged Lid and Drainage Slots, Polycarbonate (PC), 81 place (9 x 9)
SDP
Catalog No. 503028629
Encompass_Preferred
Cryogenic Boxes with Hinged Lid New ultra-low temperature storage boxes are designed to hold 1.2mL to 2.0mL cryogenic storage vials. An excellent and longlasting replacement for cardboard boxes, they are fully autoclavable and withstand temperatures ranging from -196° to +121°C. They are compatible with all standard 5.25 inch (133mm) freezer racks. Clear lids are hinged and have printed indexing for sample identification purposes. Polycarbonate, withstand -196°C Convenient hinged lid Includes drainage slots for LN2
Catalog No. 50-302-8629 Supplier Alkali Scientific Supplier No. MT2281
Cryogenic Boxes with Hinged Lid New ultra-low temperature storage boxes are designed to hold 1.2mL to 2.0mL cryogenic storage vials. An excellent and longlasting replacement for cardboard boxes, they are fully autoclavable and withstand temperatures ranging from -196° to +121°C. They are compatible with all standard 5.25 inch (133mm) freezer racks. Clear lids are hinged and have printed indexing for sample identification purposes. Polycarbonate, withstand -196°C Convenient hinged lid Includes drainage slots for LN2