Learn More
GA International Cryogenic Cover-Up Labels for Frozen Vials & Tubes 2" x 1" + 0.5" (wrap) / 50.8mm x 25.4mm + 12.7mm (wrap) Rectangle White + Transparent From -196°C to +100°C (-320.8°F to +212°F)
Catalog No. 5000570915
Encompass_Preferred
Blackout CryoSTUCK labels are designed for applying to already frozen vials and tubes and serve to conceal existing information. Ideal for re-labeling of frozen vials when removal of the existing label is not possible. These labels will completely obscure the existing label and block-out the information underneath. The re-labeled frozen vials can be stored back inside liquid nitrogen (-196°C/-321°F) or a freezer for long-term preservation. On vials and tubes, it is recommended that CryoSTUCK labels are applied with an overlap of at least 0.25" to ensure proper adherence to the frozen surface.Prior testing of these labels on end user's vials/tubes and that they are applied with an overlap of at least 0.25" for proper adherence is recommended.
Catalog No. 50-005-70915 Supplier GA International Supplier No. AEA3C11WH
Blackout CryoSTUCK labels are designed for applying to already frozen vials and tubes and serve to conceal existing information. Ideal for re-labeling of frozen vials when removal of the existing label is not possible. These labels will completely obscure the existing label and block-out the information underneath. The re-labeled frozen vials can be stored back inside liquid nitrogen (-196°C/-321°F) or a freezer for long-term preservation. On vials and tubes, it is recommended that CryoSTUCK labels are applied with an overlap of at least 0.25" to ensure proper adherence to the frozen surface.Prior testing of these labels on end user's vials/tubes and that they are applied with an overlap of at least 0.25" for proper adherence is recommended.