Learn More
Taylor Wharton CRYOGENIC DEWAR 25 LITER
Catalog No. NC0021990
Encompass
25LD cryogenic Dewar designed for storing and dispensing small amounts of liquid nitrogen. Optional equipment includes liquid withdrawal device, tipping stand, dippers and roller base.
Catalog No. NC0021990 Supplier Taylor Wharton Supplier No. 25LD
25LD cryogenic Dewar designed for storing and dispensing small amounts of liquid nitrogen. Optional equipment includes liquid withdrawal device, tipping stand, dippers and roller base.