Learn More
PHCbi Cryogenic Freezer
Catalog No. 502720063
Encompass_Preferred
PHCbi brand 8.2 cu.ft (231 L) mechanically refirgeration cryogenic freezer deploys a reliable, high performance, hybrid cascade refrigeration system. Model does not require LN2 as a primary cooling medium and stores up to 165 2: boxes or 105 3" boxesTemperature Range'-150° C
Catalog No. 50-272-0063 Supplier PHCbi Supplier No. MDFC2156VANCPA
Add to Cart
Add to Cart
Request a Quote
PHCbi brand 8.2 cu.ft (231 L) mechanically refirgeration cryogenic freezer deploys a reliable, high performance, hybrid cascade refrigeration system. Model does not require LN2 as a primary cooling medium and stores up to 165 2: boxes or 105 3" boxesTemperature Range'-150° C