Learn More
Uline Cryogenic Gloves - Large
Catalog No. NC1709551
Encompass
For protection against dry ice and ultra cold environments down to -260¿F. Lightweight and waterproof.
This item is not returnable. View return policy
Catalog No. NC1709551 Supplier Uline Supplier No. S21621L
For protection against dry ice and ultra cold environments down to -260¿F. Lightweight and waterproof.