Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Cryogenic Kit with SaferGrip™ Elbow Length Gloves
SDP

Catalog No. 17010886 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-010-886 X-Large
17-010-885 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17-010-886 Supplier National Safety Apparel™ Supplier No. KITCR003XLEL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cryogenic glove kit includes SaferGrip™ silicone coated elbow length cryogenic gloves, faceshield unit with ratchet headgear, Teflon™ laminated nylon apron, faceshield bag.

  • 3M™ Scotchlite™ Thinsulate™ liner
  • Waterproof and breathable membrane for additional splash protection
  • 100% Waterproof Liner
  • SaferGrip™ silicone coated material in the palm for enhanced grip and durability

Specifications

Certifications/Compliance (Europe) EN 511 Contact Cold Performance Level 2, EN 511 Convective Cold Performance Level 3
Color Blue
Exterior Finish Smooth
Hand Ambidextrous
Material Nylon
Product Type Cryogenic Gloves
Size X-Large
For Use With (Application) Cryogenic Liquids and Gases
Lining Lined
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.