Learn More
Labsource Inc CRYOGENIC LABELS 'CRYOBABIES'
SDP
Catalog No. NC9066164
Encompass
Cryo-Tag, Made of Polyvinyl and a Strong Adhesive that Adheres to Plastics, Metals and Glass, Withstands Temperatures of -196 to 150 deg. C and Resists Most Detergents, Oils, Organic Solvents and Caustic Agents, Size 0.94 x 0.5 in.
Catalog No. NC9066164 Supplier Labsource Inc Supplier No. L60311PK
Cryo-Tag, Made of Polyvinyl and a Strong Adhesive that Adheres to Plastics, Metals and Glass, Withstands Temperatures of -196 to 150 deg. C and Resists Most Detergents, Oils, Organic Solvents and Caustic Agents, Size 0.94 x 0.5 in.