Learn More
Electron Microscopy Sciences Cryogenic Cryostick Labels, 0.5" x 1.5"
SDP
Catalog No. 5032206
Encompass_Preferred
These labels are made from top quality materials and are designed specially for cryogenic use. For temperatures of -80 deg C. Ideal for cryotubes and are best used with thermal transfer printers or cryo marker pens. Available in a wide range of sizes on sheets or rolls. Not recommend for laser printing.
Size: 0.5" x 1.5"
Use: -80°C
Catalog No. 50-322-06 Supplier Electron Microscopy Sciences Supplier No. 7702612
These labels are made from top quality materials and are designed specially for cryogenic use. For temperatures of -80 deg C. Ideal for cryotubes and are best used with thermal transfer printers or cryo marker pens. Available in a wide range of sizes on sheets or rolls. Not recommend for laser printing.
Size: 0.5" x 1.5"
Use: -80°C