Learn More
GA International Cryogenic Labels for Direct Thermal Barcode Printers 1" x 0.5" / 25.4mm x 12.7mm Rectangle White From -196°C to +93°C (-321°F to +199°F)
Catalog No. NC3911087
Encompass_Preferred
These cryogenic barcode labels are specially designed for printing with direct thermal printers. The thermoplastic waterproof labels are specifically designed to permanently adhere to vials, microcentrifuge tubes, microtubes, microplates, glass tubes, freezer boxes, plastic bags, and other specimen containers that will undergo long-term cryogenic and deep-freeze storage. These labels have a strong affinity for glass, polypropylene, polyethylene, and other plastic surfaces. The cryogenic labels will remain adhered to tubes through multiple freeze-thaw cycles without cracking or peeling off. These direct thermal cryogenic labels are compatible with most cryogenic storage systems without any hindrance or risk of being scraped off. These labels do not require a self-laminate to work.
Catalog No. NC3911087 Supplier GA International Supplier No. DFPC7C11WH
These cryogenic barcode labels are specially designed for printing with direct thermal printers. The thermoplastic waterproof labels are specifically designed to permanently adhere to vials, microcentrifuge tubes, microtubes, microplates, glass tubes, freezer boxes, plastic bags, and other specimen containers that will undergo long-term cryogenic and deep-freeze storage. These labels have a strong affinity for glass, polypropylene, polyethylene, and other plastic surfaces. The cryogenic labels will remain adhered to tubes through multiple freeze-thaw cycles without cracking or peeling off. These direct thermal cryogenic labels are compatible with most cryogenic storage systems without any hindrance or risk of being scraped off. These labels do not require a self-laminate to work.