Learn More
GA International Cryogenic Labels for Direct Thermal Barcode Printers (Patent Pending) 0.94" x 0.5" + 0.437" (circle) / 23.9mm x 12.7mm + 11.1mm (circle) Rectangle + Circle White From -196°C to +93°C (-321°F to +199°F)
Catalog No. 5000571037
Encompass_Preferred
These are compatible with most major brands of direct thermal printers. These thermoplastic waterproof labels are specifically designed to adhere to vials, microcentrifuge tubes, microtubes, glass tubes, and other specimen containers that will undergo long-term cryogenic and deep-freeze storage. With SnapPEEL™ technology and a unique connected circle-and-rectangle design, each label allows the cap and side of vials to be labeled seamlessly in one single action. This eliminates the need for separate circle and rectangle labels, significantly improving efficiency and reducing unnecessary contact with the adhesive.
Catalog No. 50-005-71037 Supplier GA International Supplier No. DFPC560SBC12WH
These are compatible with most major brands of direct thermal printers. These thermoplastic waterproof labels are specifically designed to adhere to vials, microcentrifuge tubes, microtubes, glass tubes, and other specimen containers that will undergo long-term cryogenic and deep-freeze storage. With SnapPEEL™ technology and a unique connected circle-and-rectangle design, each label allows the cap and side of vials to be labeled seamlessly in one single action. This eliminates the need for separate circle and rectangle labels, significantly improving efficiency and reducing unnecessary contact with the adhesive.