Learn More
Electron Microscopy Sciences Cryogenic Durastick Labels, 0.5" x 0.9"
SDP
Catalog No. 5032199
Encompass_Preferred
These labels are made from top quality materials and are designed specially for cryogenic use. For Temperature of -40 deg C and below, these labels feature a smudge resistant surface. They are available in rolls for thermal transfer printing, all accept handwriten notations with cryo markers.
Size: 0.5" x 0.9"
Use: -40°C and below
Catalog No. 50-321-99 Supplier Electron Microscopy Sciences Supplier No. 7702510
These labels are made from top quality materials and are designed specially for cryogenic use. For Temperature of -40 deg C and below, these labels feature a smudge resistant surface. They are available in rolls for thermal transfer printing, all accept handwriten notations with cryo markers.
Size: 0.5" x 0.9"
Use: -40°C and below