Learn More
GA International Cryogenic Labels for Inkjet Printers 0.94" x 0.5" + 0.437" (circle) / 23.9mm x 12.7mm + 11.1mm (circle) Rectangle + Circle White From -196°C to +121°C (-321°F to +250°F)
Catalog No. 5000572675
Encompass_Preferred
Cryo-JetTAG™ cryogenic inkjet printable labels are ideal for identifying cryo vials, tubes, plastic bags, as well as cardboard and polycarbonate freezer boxes with eye-catching color logos, graphics, images, and data. These cryo labels are designed to withstand prolonged storage in cryogenic conditions, including liquid nitrogen (-196°C/-321°F) and laboratory freezers (-80°C/-112°F). Moreover, these inkjet labels also provide resistance to elevated dry heat and standard steam autoclaves, making them a good choice for use in scientific laboratories.The obtained printout has an impressive resistance to chemicals such as alcohol, acetone, and xylene, as well as to wiping and rubbing with alcohol sanitizing wipes.Various models of roll format inkjet printers can be used for Cryo-JetTAG labels according to the size of the label; These labels also can be written on with ballpoint pens, cryogenic markers, and ink stamped without smudging.
Catalog No. 50-005-72675 Supplier GA International Supplier No. CIJ52NOTC22WH
Cryo-JetTAG™ cryogenic inkjet printable labels are ideal for identifying cryo vials, tubes, plastic bags, as well as cardboard and polycarbonate freezer boxes with eye-catching color logos, graphics, images, and data. These cryo labels are designed to withstand prolonged storage in cryogenic conditions, including liquid nitrogen (-196°C/-321°F) and laboratory freezers (-80°C/-112°F). Moreover, these inkjet labels also provide resistance to elevated dry heat and standard steam autoclaves, making them a good choice for use in scientific laboratories.The obtained printout has an impressive resistance to chemicals such as alcohol, acetone, and xylene, as well as to wiping and rubbing with alcohol sanitizing wipes.Various models of roll format inkjet printers can be used for Cryo-JetTAG labels according to the size of the label; These labels also can be written on with ballpoint pens, cryogenic markers, and ink stamped without smudging.