Learn More
GA International Cryogenic Labels for Inkjet Printers 0.94" x 0.5" / 23.9mm x 12.7mm Rectangle White From -196°C to +121°C (-321°F to +250°F)
Catalog No. 5000572322
Encompass_Preferred
Cryo-JetTAG™ inkjet printable cryogenic labels are ideal for identifying cryo vials, tubes, plastic bags, as well as cardboard and polycarbonate freezer boxes destinated for cryo storage in liquid nitrogen (-196°C/-321°F) and lab freezers (-80°C/-112°F) with eye-catching color logos, graphics, images, and data. Moreover, these durable labels also resist dry heat and standard steam autoclave sterilization cycles, perfect for a variety of uses in scientific laboratories.The obtained printout offers an impressive resistance to harsh chemicals, such as alcohol, acetone, and xylene, as well as to wiping and rubbing with alcohol sanitizing wipes.Sheet format Cryo-JetTAG labels can be printed with most standard desktop inkjet printers; They can also be written on with ballpoint pens, cryogenic markers, and ink stamped without smudging.
Catalog No. 50-005-72322 Supplier GA International Supplier No. CIJSA12WH
Cryo-JetTAG™ inkjet printable cryogenic labels are ideal for identifying cryo vials, tubes, plastic bags, as well as cardboard and polycarbonate freezer boxes destinated for cryo storage in liquid nitrogen (-196°C/-321°F) and lab freezers (-80°C/-112°F) with eye-catching color logos, graphics, images, and data. Moreover, these durable labels also resist dry heat and standard steam autoclave sterilization cycles, perfect for a variety of uses in scientific laboratories.The obtained printout offers an impressive resistance to harsh chemicals, such as alcohol, acetone, and xylene, as well as to wiping and rubbing with alcohol sanitizing wipes.Sheet format Cryo-JetTAG labels can be printed with most standard desktop inkjet printers; They can also be written on with ballpoint pens, cryogenic markers, and ink stamped without smudging.