Learn More
GA International Cryogenic Labels for Laser Printers 1.28" x 0.5" / 32.5mm x 12.7mm Rectangle Lavender From -196°C to +120°C (-321°F to +248°F)
Catalog No. 5000571309
Encompass_Preferred
Cryo-LazrTAG™ thermoplastic labels are built to perform in laboratory environments. Their advanced cryo-adhesive ensures strong adhesion to various surfaces such as vials, microtubes, and polycarbonate freezer boxes stored in ultra-low temperature freezers (-80°C / -112°F) and liquid nitrogen (-196°C/-321°F). They can also withstand temperatures up to +120°C/+248°F (dry heat). These waterproof, durable labels are supplied in US letter format, allowing for easy, jam-free printing with standard desktop laser printers. Resistant to wear, handling, and mild laboratory chemicals, including swabbing with alcohols (isopropanol, ethanol) and surface sanitizing wipes, they deliver consistent reliability in demanding lab settings.
Catalog No. 50-005-71309 Supplier GA International Supplier No. CLA23LA
Cryo-LazrTAG™ thermoplastic labels are built to perform in laboratory environments. Their advanced cryo-adhesive ensures strong adhesion to various surfaces such as vials, microtubes, and polycarbonate freezer boxes stored in ultra-low temperature freezers (-80°C / -112°F) and liquid nitrogen (-196°C/-321°F). They can also withstand temperatures up to +120°C/+248°F (dry heat). These waterproof, durable labels are supplied in US letter format, allowing for easy, jam-free printing with standard desktop laser printers. Resistant to wear, handling, and mild laboratory chemicals, including swabbing with alcohols (isopropanol, ethanol) and surface sanitizing wipes, they deliver consistent reliability in demanding lab settings.