Learn More
Electron Microscopy Sciences Cryogenic Liqui-Stick Labels, 0.75" x 0.9"
SDP
Catalog No. 5032212
Encompass_Preferred
These labels are made from top quality materials and are designed specially for cryogenic use. For temperatures of -195 deg C. Synthetic surface accepts crisp bar codes from thermal transfer printers or handwritten notations cryo markers. These labels are not recommended for use with laser printers.
Size: 0.75" x 0.9"
Use: -195°C
Catalog No. 50-322-12 Supplier Electron Microscopy Sciences Supplier No. 7702715
These labels are made from top quality materials and are designed specially for cryogenic use. For temperatures of -195 deg C. Synthetic surface accepts crisp bar codes from thermal transfer printers or handwritten notations cryo markers. These labels are not recommended for use with laser printers.
Size: 0.75" x 0.9"
Use: -195°C