Learn More
Alkali Scientific ASI™ Cryogenic Labels Rolls, 9.5mm Dots for 0.5-1.5mL, Blue, 1000/Case
SDP
Catalog No. 503027253
Encompass_Preferred
We offer Cryogenic Labels Rolls 9.5mm Dots for 0.5-1.5ml that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).
Catalog No. 50-302-7253 Supplier Alkali Scientific Supplier No. LCD38B
Add to Cart
Add to Cart
We offer Cryogenic Labels Rolls 9.5mm Dots for 0.5-1.5ml that use a special adhesive designed exactly for vials, microtubes, microplates, glass tubes, freezer boxes, plastic bags and other containers. Our cryogenic storage labels will stick in temperatures ranging from 194° F (90°C) to as low as -94°F (-70°C).