Learn More
GA International Cryogenic Labels for the Identification of IVF Straws 1" x 1" / 25.4mm x 25.4mm Rectangle Pink tab From -196°C to +80°C (-321°F to +176°F)
Catalog No. 5000571430
Encompass_Preferred
Self-laminating cryogenic labels for 0.25 ml and 0.5 ml IVF straws used in assisted reproductive technology (ART) clinics and laboratories, as well as animal breeding and biotechnology companies. Designed for short and long-term cryo-preservation under cryogenic laboratory conditions, including liquid and vapor phase liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers, and dry ice transportation. Printable with thermal-transfer barcode printers for secure sample identification and traceability.Volatile Organic Compound (VOC) testing results for Cryo-StrawTAG™ labels are available and can be provided upon request in order to determine the suitability of this product for your application.
Catalog No. 50-005-71430 Supplier GA International Supplier No. CST29NPNOT0.25C31PI
Add to Cart
Add to Cart
Self-laminating cryogenic labels for 0.25 ml and 0.5 ml IVF straws used in assisted reproductive technology (ART) clinics and laboratories, as well as animal breeding and biotechnology companies. Designed for short and long-term cryo-preservation under cryogenic laboratory conditions, including liquid and vapor phase liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers, and dry ice transportation. Printable with thermal-transfer barcode printers for secure sample identification and traceability.Volatile Organic Compound (VOC) testing results for Cryo-StrawTAG™ labels are available and can be provided upon request in order to determine the suitability of this product for your application.