Learn More
GA International Cryogenic Labels for the Identification of IVF Straws 1" x 1" / 25.4mm x 25.4mm Rectangle Yellow tab From -196°C to +80°C (-321°F to +176°F)
Catalog No. 5000572369
Encompass_Preferred
Self-laminating cryogenic labels for 0.25 ml and 0.5 ml IVF straws used in assisted reproductive technology (ART) clinics and laboratories, as well as animal breeding and biotechnology companies. Designed for short and long-term cryo-preservation under cryogenic laboratory conditions, including liquid and vapor phase liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers, and dry ice transportation. Printable with thermal-transfer barcode printers for secure sample identification and traceability.Volatile Organic Compound (VOC) testing results for Cryo-StrawTAG™ labels are available and can be provided upon request in order to determine the suitability of this product for your application.
Catalog No. 50-005-72369 Supplier GA International Supplier No. CST29NPNOT0.25C11YE
Self-laminating cryogenic labels for 0.25 ml and 0.5 ml IVF straws used in assisted reproductive technology (ART) clinics and laboratories, as well as animal breeding and biotechnology companies. Designed for short and long-term cryo-preservation under cryogenic laboratory conditions, including liquid and vapor phase liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers, and dry ice transportation. Printable with thermal-transfer barcode printers for secure sample identification and traceability.Volatile Organic Compound (VOC) testing results for Cryo-StrawTAG™ labels are available and can be provided upon request in order to determine the suitability of this product for your application.