Learn More
GA International Cryogenic Labels (Writable) 0.5" / 12.7mm Circle Red From -196°C to +110°C (-321°F to +230°F)
Catalog No. 5000571285
Encompass_Preferred
These small diameter round cryo labels for microtube tops are made with a strong adhesive ideal for liquid nitrogen storage (Subject to prior verification of compatibility of labels and intended vial cap material in Liquid Nitrogen), -80°C freezers and dry ice. Specially designed to comfortably fit cryovials and Eppendorf tube caps. These waterproof deep-freeze dot labels are supplied in roll format and are available in 15 individual colors as well as packs of 15 assorted colors for color-coding and rapid identification.
Catalog No. 50-005-71285 Supplier GA International Supplier No. JTRA13RE
These small diameter round cryo labels for microtube tops are made with a strong adhesive ideal for liquid nitrogen storage (Subject to prior verification of compatibility of labels and intended vial cap material in Liquid Nitrogen), -80°C freezers and dry ice. Specially designed to comfortably fit cryovials and Eppendorf tube caps. These waterproof deep-freeze dot labels are supplied in roll format and are available in 15 individual colors as well as packs of 15 assorted colors for color-coding and rapid identification.