Learn More
GA International Cryogenic Labels (Writable) 1.02" x 0.4" / 25.9mm x 10.2mm Rectangle Lemon From -196°C to +110°C (-321°F to +230°F)
Catalog No. 5000571542
Encompass_Preferred
These writable cryogenic rectangle labels for cryo vials and conical bottom microtubes are made with a strong adhesive ideal for liquid nitrogen*, -80°C freezers, and dry ice storage and transportation. Easily writable with permanent cryogenic markers, these ultra-low temperature labels are supplied in roll format and are available in 15 individual colors as well as packs of 15 assorted colors for color coding and rapid identification.*Subject to verification of compatibility of labels and intended vial cap material in Liquid Nitrogen storage
Catalog No. 50-005-71542 Supplier GA International Supplier No. JTRA2610LE
These writable cryogenic rectangle labels for cryo vials and conical bottom microtubes are made with a strong adhesive ideal for liquid nitrogen*, -80°C freezers, and dry ice storage and transportation. Easily writable with permanent cryogenic markers, these ultra-low temperature labels are supplied in roll format and are available in 15 individual colors as well as packs of 15 assorted colors for color coding and rapid identification.*Subject to verification of compatibility of labels and intended vial cap material in Liquid Nitrogen storage