Learn More
Diversified Biotech Inc CRYOGENIC LBL 1.05X.50 2000/RL
Catalog No. 50589634
Encompass
Label, Cryogenic Freezer Label -196C, Thermal Transfer Printable, Cryo-Tags, Thermal Transfer Rolls for Cryogenic Storage, Liquid and Vapor phase Nitrogen, Solvent Resistent, 1.05 in. x 0.50 in.,27mm x 13mm, White, Gloss Finish, 2,000/RL
Catalog No. 50-589-634 Supplier Diversified Biotech Inc Supplier No. CRTH2000
Label, Cryogenic Freezer Label -196C, Thermal Transfer Printable, Cryo-Tags, Thermal Transfer Rolls for Cryogenic Storage, Liquid and Vapor phase Nitrogen, Solvent Resistent, 1.05 in. x 0.50 in.,27mm x 13mm, White, Gloss Finish, 2,000/RL