Learn More
Andwin Scientific Laser Cryo-Tags, 1.125 x 1.75" (28x44mm), 840/pk
SDP
Catalog No. NC1659366
Encompass_Preferred
Our patented Cryo-Babies and Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage. Cryogenic labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks.
Catalog No. NC1659366 Supplier Andwin Scientific Supplier No. LCRY1300
Our patented Cryo-Babies and Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage. Cryogenic labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks.