Learn More
Andwin Scientific Cryo-Clear Laser Sheet Label, 1.28x0.5inch, 1700/pk
SDP
Catalog No. NC1660842
Encompass_Preferred
Cryo-Clear Laser Sheet Label, 1.28x0.5inch, 1700/pk, Temperature Resistant-196 degree C to 70 degree C. Remain fully adhered down to -196 degree C. Contents of vials beneath labels remain visible.
Catalog No. NC1660842 Supplier Andwin Scientific Supplier No. CLEAR170
Cryo-Clear Laser Sheet Label, 1.28x0.5inch, 1700/pk, Temperature Resistant-196 degree C to 70 degree C. Remain fully adhered down to -196 degree C. Contents of vials beneath labels remain visible.