Learn More
Diversified Biotech Inc Cryo-Tags 1.50 x 0.75" 1,000/roll
Catalog No. NC1691556
Encompass
Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage. Cryogenic labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks. Cryo-Tags also withstand flash/snap freezing in liquid nitrogen.
Catalog No. NC1691556 Supplier Diversified Biotech Inc Supplier No. CRY2000
Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage. Cryogenic labels are chemically inert with a solvent resistant adhesive that will adhere to croygenic vials, microtubes, test tubes, glass vials, beakers and metal racks. Cryo-Tags also withstand flash/snap freezing in liquid nitrogen.