Learn More
Andwin Scientific Laser Cryo-Babies 0.94 x 0.50", Red, 2380/pk
SDP
Catalog No. NC1660216
Encompass_Preferred
Our patented Cryo-Babies and Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage.
Catalog No. NC1660216 Supplier Andwin Scientific Supplier No. LCRY2380R
Our patented Cryo-Babies and Cryo-Tags withstand both long term cryogenic storage conditions and conventional freezer storage.