Learn More
Andwin Scientific DT Cryo-Spots. 1/2" diam., 4,000/roll, Green
SDP
Catalog No. NC1660838
Encompass_Preferred
Direct Thermal Cryo-Tags are formatted to easily work with the Dymo Labelwriter. Quickly label your samples with sharp text, clear barcodes or a combination of both!
Catalog No. NC1660838 Supplier Andwin Scientific Supplier No. DTCR4000G
Direct Thermal Cryo-Tags are formatted to easily work with the Dymo Labelwriter. Quickly label your samples with sharp text, clear barcodes or a combination of both!