Learn More
Mitegen Waterproof Cryo-Gloves, Mid-Arm Length, Large Size, Blue, For LN2 Splashes & Spills, -196°C Protection
SDP
Catalog No. NC3245526
Encompass
These Waterproof Cryo-Gloves (Mid-Arm, Large, Blue) provide superior hand protection for laboratory professionals working with liquid nitrogen and other cryogenic fluids. Featuring a thin, seamless 100% waterproof liner, these gloves offer robust defense against spills and splashes. Designed for ultra-cold applications down to -196°C (-320°F), their state-of-the-art multi-layered construction ensures maximum thermal and splash protection, while maintaining essential flexibility and dexterity. A high-performance thermal inner lining actively wicks moisture away from hands, providing sustained comfort over extended periods. These gloves are safety certified to meet EU standards (CE Cat. III: EN 511, EN 388, EN 420). Ideal for dispensing/transferring cryogenic liquids, removing samples from cryogenic liquids, or any tasks involving open cryogenic containers with the chance of exposure to liquids or splashes.
Catalog No. NC3245526 Supplier Mitegen Supplier No. TSMALWP
These Waterproof Cryo-Gloves (Mid-Arm, Large, Blue) provide superior hand protection for laboratory professionals working with liquid nitrogen and other cryogenic fluids. Featuring a thin, seamless 100% waterproof liner, these gloves offer robust defense against spills and splashes. Designed for ultra-cold applications down to -196°C (-320°F), their state-of-the-art multi-layered construction ensures maximum thermal and splash protection, while maintaining essential flexibility and dexterity. A high-performance thermal inner lining actively wicks moisture away from hands, providing sustained comfort over extended periods. These gloves are safety certified to meet EU standards (CE Cat. III: EN 511, EN 388, EN 420). Ideal for dispensing/transferring cryogenic liquids, removing samples from cryogenic liquids, or any tasks involving open cryogenic containers with the chance of exposure to liquids or splashes.