Learn More
Research Products International Corp Cryogenic Storage Box, Hinged Lid, 100 Tube Capacity, Assorted Colors, 5 per Case
SDP
Catalog No. 50476457
Encompass
Durable polypropylene storage boxes withstand repeated low temperature storage. Plasastic boxes will not deterioate or saturate like cardboard boxes. Hinged lid provides convenient access to sample tubes. Available in assorted colors (green, red, blue, orange, yellow). Accommodates all hinged and screw cap 1.5-2.0ml micro-tubes.
Catalog No. 50-476-457 Supplier Research Products International Corp Supplier No. 145634
Durable polypropylene storage boxes withstand repeated low temperature storage. Plasastic boxes will not deterioate or saturate like cardboard boxes. Hinged lid provides convenient access to sample tubes. Available in assorted colors (green, red, blue, orange, yellow). Accommodates all hinged and screw cap 1.5-2.0ml micro-tubes.