Learn More
Sigma Aldrich Fine Chemicals Biosciences Cryogenic storage rack sta
Catalog No. 502908994
Encompass
Racks hold CryoBoxes securely but separately allowing fast retrieval and immediate return to freezer. The vertical racks are best for use in chest freezers horizontal racks for upright freezers.
Catalog No. 50-290-8994 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. R13882EA
Racks hold CryoBoxes securely but separately allowing fast retrieval and immediate return to freezer. The vertical racks are best for use in chest freezers horizontal racks for upright freezers.