Learn More
GA International Cryogenic Tape for Frozen Containers: with GatorCUT™ Technology (Patented) 1" x 50' / 25.4mm x 15m Blue 1" / 25.4mm From -196°C to +100°C (-321°F to +212°F) / Minimum Application Temperature: -80°C (-112°F)
Catalog No. NC3240997
Encompass_Preferred
This cryogenic tape is especially designed to be applied directly, and rapidly adhere, to frozen cryo vials, tubes, polycarbonate cryo boxes, plastic and cardboard freezer boxes, and other cryogenic containers which are difficult to label under deep-freeze conditions (-80°C/-112°F). Featuring GA international’s patented GatorCUT™ technology to effortlessly hand tear as little or as much needed without needing a tape dispenser or scissors. Designed for long-term preservation in liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers, and dry ice transportation, they can also withstand water exposure such as thawing in water baths, as well as temperatures up to +100°C (212°F). Can be easily written on with our permanent cryo-markers.
Catalog No. NC3240997 Supplier GA International Supplier No. TFNG25SBC150BL
This cryogenic tape is especially designed to be applied directly, and rapidly adhere, to frozen cryo vials, tubes, polycarbonate cryo boxes, plastic and cardboard freezer boxes, and other cryogenic containers which are difficult to label under deep-freeze conditions (-80°C/-112°F). Featuring GA international’s patented GatorCUT™ technology to effortlessly hand tear as little or as much needed without needing a tape dispenser or scissors. Designed for long-term preservation in liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers, and dry ice transportation, they can also withstand water exposure such as thawing in water baths, as well as temperatures up to +100°C (212°F). Can be easily written on with our permanent cryo-markers.