Learn More
GA International Cryogenic Tape with GatorCUT™ Technology (Patented) 1" x 50' / 25.4mm x 15m Red 1" / 25.4mm From -196°C to +110°C (-321°F to +230°F)
Catalog No. NC3240992
Encompass_Preferred
Continuous writable tape for cryogenic use on cryo/freezer boxes, bottles, tubes and vials. Featuring our innovative, patent-pending GatorCUT™ technology that allows the tape to be effortlessly hand-cut without the use of scissors or dispensers. Conveniently portable for use whenever and wherever needed, and coated with our extra strong CryoMax™ adhesive for secure, long-term container identification. Waterproof, resistant to immersion in liquid nitrogen (-196°C/-321°F), transport on dry ice, multiple freeze-thaw cycles, and other harsh laboratory environments. Available in a variety of colors for easy color coding and quick identification of plastic and freezer-grade glass containers.
Catalog No. NC3240992 Supplier GA International Supplier No. TJTG25SBC150RE
Continuous writable tape for cryogenic use on cryo/freezer boxes, bottles, tubes and vials. Featuring our innovative, patent-pending GatorCUT™ technology that allows the tape to be effortlessly hand-cut without the use of scissors or dispensers. Conveniently portable for use whenever and wherever needed, and coated with our extra strong CryoMax™ adhesive for secure, long-term container identification. Waterproof, resistant to immersion in liquid nitrogen (-196°C/-321°F), transport on dry ice, multiple freeze-thaw cycles, and other harsh laboratory environments. Available in a variety of colors for easy color coding and quick identification of plastic and freezer-grade glass containers.