Learn More
GA International Cryogenic Tape for Frozen Containers: with GatorCUT™ Technology (Patented) 1" x 50' / 25.4mm x 15m Yellow 1" / 25.4mm From -196°C to +100°C (-321°F to +212°F) / Minimum Application Temperature: -80°C (-112°F)
Catalog No. NC3457133
Encompass_Preferred
This cryogenic tape is especially designed to be applied directly, and rapidly adhere, to frozen cryo vials, tubes, polycarbonate cryo boxes, plastic and cardboard freezer boxes, and other cryogenic containers which are difficult to label under deep-freeze conditions (-80°C/-112°F). Featuring GA international’s patented GatorCUT™ technology to effortlessly hand tear as little or as much needed without needing a tape dispenser or scissors. Designed for long-term preservation in liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers, and dry ice transportation, they can also withstand water exposure such as thawing in water baths, as well as temperatures up to +100°C (212°F). Can be easily written on with our permanent cryo-markers.
Catalog No. NC3457133 Supplier GA International Supplier No. TFNG25SBC150YE
This cryogenic tape is especially designed to be applied directly, and rapidly adhere, to frozen cryo vials, tubes, polycarbonate cryo boxes, plastic and cardboard freezer boxes, and other cryogenic containers which are difficult to label under deep-freeze conditions (-80°C/-112°F). Featuring GA international’s patented GatorCUT™ technology to effortlessly hand tear as little or as much needed without needing a tape dispenser or scissors. Designed for long-term preservation in liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers, and dry ice transportation, they can also withstand water exposure such as thawing in water baths, as well as temperatures up to +100°C (212°F). Can be easily written on with our permanent cryo-markers.