Learn More
GA International Cryogenic Tape for Metal Racks 1" x 50' / 25.4mm x 15m White 1" / 25.4mm From -196°C to +100°C (-321°F to +212°F)
Catalog No. NC3806107
Encompass_Preferred
Cryogenic tape for identifying various metal containers for long-term cryogenic storage, as low as -196°C (-321°F). This cryo tape can be used to identify stainless-steel and aluminum racks, X-ray cassettes, film cans, and more. The tape should be applied at room temperature before being placed in liquid nitrogen or ultra-low temperature freezers for subsequent storage. Writable with our permanent cryo markers, this tape for metal racks can withstand storage in liquid nitrogen, transportation on dry ice, multiple freeze-thaw cycles, as well as gamma irradiation (up to 50 KGy).
Catalog No. NC3806107 Supplier GA International Supplier No. TWB25SBC150WH
Cryogenic tape for identifying various metal containers for long-term cryogenic storage, as low as -196°C (-321°F). This cryo tape can be used to identify stainless-steel and aluminum racks, X-ray cassettes, film cans, and more. The tape should be applied at room temperature before being placed in liquid nitrogen or ultra-low temperature freezers for subsequent storage. Writable with our permanent cryo markers, this tape for metal racks can withstand storage in liquid nitrogen, transportation on dry ice, multiple freeze-thaw cycles, as well as gamma irradiation (up to 50 KGy).