Learn More
GA International Cryogenic Thermal-Transfer Labels for Frozen Vials & Tubes 2.75" x 1.1875" + 1.25" (wrap) / 69.9mm x 30.2mm + 31.8mm (wrap) Rectangle White + Transparent From -196°C to +110°C (-321°F to +230°F)
Catalog No. 5000571457
Encompass_Preferred
Labeling of frozen vials is challenging due to extremely low temperatures, frost and moisture accumulated on the surface of these cryogenic containers. CryoSTUCK labels provide an ideal solution for bio-banks and bio-repositories that need relabeling of already frozen samples inside liquid nitrogen tanks and freezers. These cryogenic labels are available in roll format for thermal-transfer printing. On vials and tubes, it is recommended that CryoSTUCK labels are applied with an overlap of at least 0.25” to ensure proper adherence to the frozen surface.
Catalog No. 50-005-71457 Supplier GA International Supplier No. FSA334C31WH
Labeling of frozen vials is challenging due to extremely low temperatures, frost and moisture accumulated on the surface of these cryogenic containers. CryoSTUCK labels provide an ideal solution for bio-banks and bio-repositories that need relabeling of already frozen samples inside liquid nitrogen tanks and freezers. These cryogenic labels are available in roll format for thermal-transfer printing. On vials and tubes, it is recommended that CryoSTUCK labels are applied with an overlap of at least 0.25” to ensure proper adherence to the frozen surface.