Learn More
GA International Cryogenic Thermal-Transfer Labels for Metal Racks 3" x 2" / 76.2mm x 50.8mm Rectangle White From -196°C to +100°C (-320.8°F to +212°F)
Catalog No. 5000572165
Encompass_Preferred
Cryogenic labels for identification of metal surfaces in liquid nitrogen Dewars, such as aluminum cryogenic vial canes, metal racks for cryo and freezer boxes, as well as aluminum cassettes and metal sleeves. Specifically designed for easy marking and identification inside Dewar-type liquid-nitrogen tanks and laboratory freezers. These specialty labels are made from a cryogenic material that has been tested and approved for applying to stainless steel and aluminum with subsequent immersion and storage inside liquid phase and vapor phase liquid nitrogen. These MetaliTAG™ cryo labels can be printed with all major brands of thermal-transfer printers and can be provided in different colors and configurations upon request.
Catalog No. 50-005-72165 Supplier GA International Supplier No. AWB56C11WH
Cryogenic labels for identification of metal surfaces in liquid nitrogen Dewars, such as aluminum cryogenic vial canes, metal racks for cryo and freezer boxes, as well as aluminum cassettes and metal sleeves. Specifically designed for easy marking and identification inside Dewar-type liquid-nitrogen tanks and laboratory freezers. These specialty labels are made from a cryogenic material that has been tested and approved for applying to stainless steel and aluminum with subsequent immersion and storage inside liquid phase and vapor phase liquid nitrogen. These MetaliTAG™ cryo labels can be printed with all major brands of thermal-transfer printers and can be provided in different colors and configurations upon request.