Learn More
GA International Cryogenic Thermal-Transfer Wrap-Around + Circle Labels 0.375" (circle) + 1" x 0.75" + 1.75" (wrap) / 9.5mm (circle) + 25.4mm x 19.1mm + 44.5mm (wrap) Rectangle + Circle Blue + Transparent From -196°C to +100°C (-321°F to +212°F)
Catalog No. 5000572398
Encompass_Preferred
Cryo-compatible, self-laminating labels that protect the printed information from alcohol, bleach, DMSO and detergents to ensure the secure identification of tubes, cryovials, and caps. Designed for long-term preservation under cryogenic laboratory conditions including liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers and dry ice, and can withstand temperatures up to +100°C (+212°F). Labels can be written on with cryo-markers, and printed with all major brands of thermal-transfer printers.
Catalog No. 50-005-72398 Supplier GA International Supplier No. JBTTA1NOTC12BL
Cryo-compatible, self-laminating labels that protect the printed information from alcohol, bleach, DMSO and detergents to ensure the secure identification of tubes, cryovials, and caps. Designed for long-term preservation under cryogenic laboratory conditions including liquid nitrogen (-196°C/-321°F), -80°C (-112°F) freezers and dry ice, and can withstand temperatures up to +100°C (+212°F). Labels can be written on with cryo-markers, and printed with all major brands of thermal-transfer printers.