Learn More
Chemglass Life Sciences Cryogenic Vial, 2.0mL, External Thread, Self-Standing, Sterile, 50/Pk, 500/Box
SDP
Catalog No. 501897759
Encompass_Preferred
Cryogenic vial with special screw cap designed without silicone ring to avoid the contamination risk. • USP grade VI Polypropylene • Autoclave at 121°C• Manufactured in clean room • Clear graduations and white marking area • Sterilized by E-Beam • Non-pyrogenic, DNase/RNase free Internal-Thread Cryogenic Vials • Used for storage in the gas phase of liquid nitrogen • Stand upright design for the bottom of the tube allows for self-standing • Optional color inserts available separately for caps
Catalog No. 50-189-7759 Supplier Chemglass Life Sciences Supplier No. CGN476202A
Cryogenic vial with special screw cap designed without silicone ring to avoid the contamination risk. • USP grade VI Polypropylene • Autoclave at 121°C• Manufactured in clean room • Clear graduations and white marking area • Sterilized by E-Beam • Non-pyrogenic, DNase/RNase free Internal-Thread Cryogenic Vials • Used for storage in the gas phase of liquid nitrogen • Stand upright design for the bottom of the tube allows for self-standing • Optional color inserts available separately for caps