Learn More
Chemglass Life Sciences Cryogenic Vial, 3.0mL, Internal Thread, Self-Standing, Sterile, 50/Pk, 250/Box
SDP
Catalog No. 501897766
Encompass_Preferred
Cryogenic vial with special screw cap designed with silicone washer for a secure seal.USP grade VI PolypropyleneAutoclave at 121°CManufactured in clean roomClear graduations and white marking areaSterilized by E-BeamNon-pyrogenic, DNase/RNase free Internal-Thread Cryogenic VialsUsed for storage in the gas phase of liquid nitrogenStand upright design for the bottom of the tube allows for self-standingOptional color inserts available separately for caps
Catalog No. 50-189-7766 Supplier Chemglass Life Sciences Supplier No. CGN4766003
Cryogenic vial with special screw cap designed with silicone washer for a secure seal.USP grade VI PolypropyleneAutoclave at 121°CManufactured in clean roomClear graduations and white marking areaSterilized by E-BeamNon-pyrogenic, DNase/RNase free Internal-Thread Cryogenic VialsUsed for storage in the gas phase of liquid nitrogenStand upright design for the bottom of the tube allows for self-standingOptional color inserts available separately for caps