Learn More
Corning™ Cryogenic Vial Grippers
Catalog No. 07210136 Shop All Corning Products
Change view
Click to view available options
Quantity:
5/Pk.
Catalog No. Quantity
07-210-136 5/Pk.
1 options
This item is not returnable. View return policy
Catalog No. 07-210-136 Supplier Corning™ Supplier No. 432136
Get Up to $2,500 in Products with Purchase
View all promotions
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Enables to easily sort or move vials maintaining sterility. Corning™ Cryogenic Vial Grippers feature unique design to grasp internal or external-thread cryogenic vials.
Cryogenic vial grippers feature a unique design to grasp internal or external-thread cryogenic vials. Grippers enable to easily sort or move vials while maintaining sterility and protecting fingers from frozen vials, dry ice and liquid nitrogen. 5 per case.
TRUSTED_SUSTAINABILITY
Specifications
Type Vial Grippers
For Use With (Application) Sorting and moving cryogenic vials
Length (Metric) Exterior 15.2 cm
Quantity 5/Pk.
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov