Learn More
Molecular Dimensions CRYOGENIC VIAL HOLDER
SDP
Catalog No. 501038569
Encompass
Cryogenic Vial Holder White polycarbonate cryogenic vial holder. Allows single handed vial manipulation. Bottom of each well interlocks with base of vial. Holds 50 vials in a 5 x 10 array.
Catalog No. 50-103-8569 Supplier Molecular Dimensions Supplier No. MD727
Cryogenic Vial Holder White polycarbonate cryogenic vial holder. Allows single handed vial manipulation. Bottom of each well interlocks with base of vial. Holds 50 vials in a 5 x 10 array.