Learn More
Biotang Inc True North* Cryogenic Vials, 50/bag, 500/cs, sterile, 2.0mL with Natural-color lids. Each vial has extra-thick walls for superior puncture and rupture protection, imprinted writing area, silicone rubber seal ring, and self-standing base.
Catalog No. 507515034 Shop All BIOTANG Inc Products
Encompass
True North* Cryogenic Vials, 50/bag, 500/cs, sterile, 2.0mL with Natural-color lids. Each vial has extra-thick walls for superior puncture and rupture protection, imprinted writing area, silicone rubber seal ring, and self-standing base.
Catalog No. 50-751-5034 Supplier Biotang Inc Supplier No. HS23202N
True North* Cryogenic Vials, 50/bag, 500/cs, sterile, 2.0mL with Natural-color lids. Each vial has extra-thick walls for superior puncture and rupture protection, imprinted writing area, silicone rubber seal ring, and self-standing base.