Learn More
Globe Scientific Cryogenic Vial Picker
Catalog No. 220101130 Shop All Globe Scientific Products
Change view
Click to view available options
Color:
Blue
Catalog No. Color
22-010-1130 Blue
1 options
This item is not returnable. View return policy
Catalog No. 22-010-1130 Supplier Globe Scientific Supplier No. 30078
Add to Cart
Add to Cart
This item is not returnable. View return policy
For safe and convenient removal of frozen vials from storage boxes
  • For safe and convenient removal of frozen vials from storage boxes
  • Simply insert picker into notch located in top of cryoclear caps and pull vial out
Specifications
Color Blue
Sterility Non-sterile