Learn More
Electron Microscopy Sciences Cryogenic Vial Rack, 7½" x 4½" x ⅞" 
SDP
Catalog No. 50996444
Encompass_Preferred
Made from durable polycarbonate. Its unique design with slotted cavities accepts vials with slotted skirts. When placed in the holes the vials become locked in position, enabling the user to attach or remove the cap with one hand. It accommodates up to 50 vials (1/2 " or 13.5 mm diameter) in a 4x10 array with alphanumeric indexing. Autoclavable at 121 degree C.
Catalog No. 50-996-444 Supplier Electron Microscopy Sciences Supplier No. 6201501
Made from durable polycarbonate. Its unique design with slotted cavities accepts vials with slotted skirts. When placed in the holes the vials become locked in position, enabling the user to attach or remove the cap with one hand. It accommodates up to 50 vials (1/2 " or 13.5 mm diameter) in a 4x10 array with alphanumeric indexing. Autoclavable at 121 degree C.